4-(1-(tert-Butoxycarbonyl)piperidin-4-yl)benzoic acid 98%

Supplier: Ambeed
Warning

A616220-10G A616220-1G A616220-250MG A616220-25G A616220-5G
77018-088EA 282.2 USD
77018-088 77018-090 77018-092 77018-094 77018-096
4-(1-(tert-Butoxycarbonyl)piperidin-4-yl)benzoic acid 98%
4-(1-(tert-Butoxycarbonyl)piperidin-4-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₄
MW: 305.36 g/mol
MDL Number: MFCD02178897
CAS Number: 149353-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR