(R)-4-Cyano-3-hydrobutyric acid ethyl ester 95%

Supplier: Ambeed
Danger

Synonyms: Ethyl 4-cyanobutanoate

A615958-100MG A615958-1G A615958-250MG A615958-5G
77018-078EA 26.53 USD
77018-078 77018-080 77018-082 77018-084
(R)-4-Cyano-3-hydrobutyric acid ethyl ester 95%
(R)-4-Cyano-3-hydrobutyric acid ethyl ester

Precaución: for Research Use Only

Formula: C₇H₁₁NO₂
MW: 141.168 g/mol
MDL Number: MFCD11040664
CAS Number: 10444-38-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR