2,4-Difluoro-5-nitroanisole 96%

Supplier: Ambeed
Warning

Synonyms: 1,5-Difluoro-2-methoxy-4-nitrobenzene

A615957-1G A615957-25G A615957-5G
77005-468EA 65.78 USD
77005-468 77005-470 77005-472
2,4-Difluoro-5-nitroanisole 96%
2,4-Difluoro-5-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₃
MW: 189.1163 g/mol
MDL Number: MFCD08689675
CAS Number: 179011-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR