5-Methoxyquinolin-8-ol 95%
Supplier: Ambeed
|
Warning
|
A615298-1G
A615298-250MG
A615298-100MG
77018-070EA
270.53
USD
77018-070
77613-910
77684-495
5-Methoxyquinolin-8-ol 95%
5-Methoxyquinolin-8-ol
Precaución: For research use only.
Formula:
C₁₀H₉NO₂ MW: 175.18 g/mol |
MDL Number:
MFCD18414704 CAS Number: 57334-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...