5-Amino-1-methyl-1H-imidazole-4-carboxamide 97%

Supplier: Ambeed
Warning

A614981-100MG A614981-1G A614981-250MG A614981-50MG
76992-174EA 29.71 USD
76992-174 76992-176 76992-178 76992-180
5-Amino-1-methyl-1H-imidazole-4-carboxamide 97%
5-Amino-1-methyl-1H-imidazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₅H₈N₄O
MW: 140.14 g/mol
MDL Number: MFCD14281882
CAS Number: 21343-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR