2-Bromo-3-nitropyridine-4-amine 97%

Supplier: Ambeed
Warning

A614758-100MG A614758-1G A614758-250MG A614758-5G
77035-220EA 20 USD
77035-220 77035-222 77035-224 77669-638
2-Bromo-3-nitropyridine-4-amine 97%
2-Bromo-3-nitropyridine-4-amine

Precaución: for Research Use Only

Formula: C₅H₄BrN₃O₂
MDL Number: MFCD11656226
CAS Number: 84487-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR