2-Fluoro-3-methoxyphenol 98%

Supplier: Ambeed
Danger

A614367-1G A614367-25G A614367-5G A614367-100G
77018-064EA 11 USD
77018-064 77018-066 77018-068 77893-238
2-Fluoro-3-methoxyphenol 98%
2-Fluoro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
MDL Number: MFCD11110186
CAS Number: 447462-87-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR