2-Chloro-6-hydroxybenzonitrile 97%

Supplier: Ambeed
Danger

A614365-10G A614365-1G A614365-5G A614365-25G
77035-214EA 70 USD
77035-214 77035-216 77035-218 77873-007
2-Chloro-6-hydroxybenzonitrile 97%
2-Chloro-6-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClNO
MDL Number: MFCD01646165
CAS Number: 89999-90-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR