3-Iodo-4-nitro-1H-indazole 95%

Supplier: Ambeed
Warning

A614228-100MG A614228-1G A614228-250MG A614228-5G
77005-458EA 18.04 USD
77005-458 77005-460 77005-462 77005-464
3-Iodo-4-nitro-1H-indazole 95%
3-Iodo-4-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄IN₃O₂
MW: 289.0300 g/mol
MDL Number: MFCD07781574
CAS Number: 885521-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR