2-Fluoro-4-morpholinobenzoic acid 97%

Supplier: Ambeed
Warning

A613904-1G A613904-250MG A613904-5G
77035-208EA 93.36 USD
77035-208 77035-210 77035-212
2-Fluoro-4-morpholinobenzoic acid 97%
2-Fluoro-4-morpholinobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₃
MW: 225.22 g/mol
MDL Number: MFCD11040390
CAS Number: 946598-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR