(2-Chloro-1,3-thiazol-5-yl)methanol 95%

Supplier: Ambeed
Warning

A613585-100MG A613585-250MG A613585-5G
77005-452EA 20.16 USD
77005-452 77005-454 77005-456
(2-Chloro-1,3-thiazol-5-yl)methanol 95%
(2-Chloro-1,3-thiazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₄H₄NOClS
MDL Number: MFCD03001277
CAS Number: 145015-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR