6-Bromo-2-chloro-3-methylquinoxaline 95%

Supplier: Ambeed
Warning

A612096-100MG A612096-1G A612096-250MG
77018-048EA 88.06 USD
77018-048 77018-050 77018-052
6-Bromo-2-chloro-3-methylquinoxaline 95%
6-Bromo-2-chloro-3-methylquinoxaline

Precaución: for Research Use Only

Formula: C₉H₆BrClN₂
MW: 257.51 g/mol
MDL Number: MFCD23162635
CAS Number: 98416-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR