(1R,4R)-2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride 96%

Supplier: Ambeed
Warning

A611999-100MG A611999-1G A611999-250MG A611999-5G
76992-152EA 15.92 USD
76992-152 76992-154 76992-156 76992-158
(1R,4R)-2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride 96%
(1R,4R)-2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO
MW: 135.59 g/mol
MDL Number: MFCD01321291
CAS Number: 601515-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR