9-Acetylanthracene 98%

Supplier: Ambeed
Warning

A611926-100G A611926-1G A611926-25G A611926-500G A611926-5G
77035-180EA 287 USD
77035-180 77035-182 77035-184 77035-186 77035-188
9-Acetylanthracene 98%
9-Acetylanthracene

Precaución: for Research Use Only

Formula: C₁₆H₁₂O
MDL Number: MFCD00001259
CAS Number: 784-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR