Methyl 3-Bromo-5-(bromomethyl)benzoate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 3-bromo-5-(bromomethyl)benzoate
A611790-10G
A611790-250MG
A611790-25G
A611790-5G
76992-144EA
843.42
USD
76992-144
76992-146
76992-148
76992-150
Methyl 3-Bromo-5-(bromomethyl)benzoate 98%
Methyl 3-Bromo-5-(bromomethyl)benzoate
Precaución: for Research Use Only
Formula:
C₉H₈Br₂O₂ MW: 307.967 g/mol |
MDL Number:
MFCD11044991 CAS Number: 877624-40-3 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...