1-Ethyl-1H-pyrazole-5-carbaldehyde 95+%

Supplier: Ambeed
Warning

A611106-1G A611106-250MG A611106-5G
77035-174EA 30 USD
77035-174 77035-176 77035-178
1-Ethyl-1H-pyrazole-5-carbaldehyde 95+%
1-Ethyl-1H-pyrazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD03130036
CAS Number: 902837-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR