Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic acid Ethyl ester, Ethyl 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylate

A609434-1G A609434-250MG A609434-5G
77035-162EA 7.43 USD
77035-162 77035-164 77035-166
Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate 98%
Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₅F₂NO₄
MW: 323.30 g/mol
MDL Number: MFCD05864419
CAS Number: 112811-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR