(R)-3-(Dimethylamino)-1-(thiophen-2-yl)propan-1-ol 95%

Supplier: Ambeed
Warning

A609098-100MG A609098-1G A609098-250MG
76992-116EA 87 USD
76992-116 76992-118 76992-120
(R)-3-(Dimethylamino)-1-(thiophen-2-yl)propan-1-ol 95%
(R)-3-(Dimethylamino)-1-(thiophen-2-yl)propan-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₅NOS
MDL Number: MFCD11558915
CAS Number: 132335-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR