1-(4-Methoxybenzyl)piperidine-2,4-dione 95%
Supplier: Ambeed
|
Warning
|
A608605-100MG
A608605-1G
A608605-250MG
76992-110EA
16.98
USD
76992-110
76992-112
76992-114
1-(4-Methoxybenzyl)piperidine-2,4-dione 95%
1-(4-Methoxybenzyl)piperidine-2,4-dione
Precaución: for Research Use Only
Formula:
C₁₃H₁₅NO₃ MW: 233.26 g/mol |
MDL Number:
MFCD17011792 CAS Number: 712353-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...