3-Aminocyclobutanol hydrochloride 98%

Supplier: Ambeed
Warning

A607886-100MG A607886-250MG A607886-5G A607886-1G
77005-442EA 19.1 USD
77005-442 77005-444 77005-446 77602-542
3-Aminocyclobutanol hydrochloride 98%
3-Aminocyclobutanol hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD12547215
CAS Number: 1036260-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR