6-Fluoro-4-iodo-1H-indole 95%

Supplier: Ambeed
Warning

A607654-100MG A607654-1G A607654-250MG
77018-018EA 38.2 USD
77018-018 77018-020 77018-022
6-Fluoro-4-iodo-1H-indole 95%
6-Fluoro-4-iodo-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅FIN
MW: 261.03 g/mol
MDL Number: MFCD07781710
CAS Number: 885520-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR