2-Methoxy-4-methylaniline hydrochloride 98%

Supplier: Ambeed
Warning

A607642-100MG A607642-250MG A607642-5G A607642-1G
77005-440EA 10 USD
77005-440 77882-430 77882-431 77882-429
2-Methoxy-4-methylaniline hydrochloride 98%
2-Methoxy-4-methylaniline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNO
MW: 173.6400 g/mol
MDL Number: MFCD09878311
CAS Number: 71288-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR