2-Bromo-6-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A607440-1G A607440-250MG A607440-5G
77035-154EA 54.11 USD
77035-154 77035-156 77035-158
2-Bromo-6-nitrobenzoic acid 95%
2-Bromo-6-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrNO₄
MW: 246.0200 g/mol
MDL Number: MFCD13688997
CAS Number: 38876-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR