N2,N2'-(Ethane-1,2-diyl)bis(N2-(3-((4,6-bis(butyl(1,2,2,6,6-pentamethylpiperidin-4-yl)amino)-1,3,5-triazin-2-yl)amino)propyl)-N4,N6-dibutyl-N4,N6-bis(1,2,2,6,6-pentamethylpiperidin-4-yl)-1,3,5-triazine-2,4,6-triamine) 99+%

Supplier: Ambeed
Danger

A607433-25G A607433-5G
76992-106EA 37 USD
76992-106 76992-108
N2,N2'-(Ethane-1,2-diyl)bis(N2-(3-((4,6-bis(butyl(1,2,2,6,6-pentamethylpiperidin-4-yl)amino)-1,3,5-triazin-2-yl)amino)propyl)-N4,N6-dibutyl-N4,N6-bis(1,2,2,6,6-pentamethylpiperidin-4-yl)-1,3,5-triazine-2,4,6-triamine) 99+%
N2,N2'-(Ethane-1,2-diyl)bis(N2-(3-((4,6-bis(butyl(1,2,2,6,6-pentamethylpiperidin-4-yl)amino)-1,3,5-triazin-2-yl)amino)propyl)-N4,N6-dibutyl-N4,N6-bis(1,2,2,6,6-pentamethylpiperidin-4-yl)-1,3,5-triazine-2,4,6-triamine)

Precaución: for Research Use Only

Formula: C₁₃₂H₂₅₀N₃₂
MW: 2285.61 g/mol
MDL Number: MFCD00134691
CAS Number: 106990-43-6
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR