(2,6-Dichlorobenzyl)triphenylphosphonium chloride 95+%
Supplier: Ambeed
|
Warning
|
A607397-1G
A607397-25G
A607397-5G
77035-148EA
16.98
USD
77035-148
77035-150
77035-152
(2,6-Dichlorobenzyl)triphenylphosphonium chloride 95+%
(2,6-Dichlorobenzyl)triphenylphosphonium chloride
Precaución: for Research Use Only
Formula:
C₂₅H₂₀Cl₃P MW: 457.78 g/mol |
MDL Number:
MFCD00061392 CAS Number: 18583-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...