3,5-Dichloro-4-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A607329-250MG A607329-5G
77005-432EA 30 USD
77005-432 77005-434
3,5-Dichloro-4-methoxybenzaldehyde 95%
3,5-Dichloro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MDL Number: MFCD04070684
CAS Number: 41727-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR