3-Chloro-4-(1H-pyrazol-1-yl)benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(1-pyrazolyl)benzaldehyde

A607234-100MG A607234-1G A607234-250MG
77035-142EA 42.44 USD
77035-142 77035-144 77035-146
3-Chloro-4-(1H-pyrazol-1-yl)benzaldehyde 98%
3-Chloro-4-(1H-pyrazol-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂O
MW: 206.6284 g/mol
MDL Number: MFCD11656795
CAS Number: 1186663-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR