5-Chloro-1H-pyrazol-3-amine 98%
Supplier: Ambeed
|
Warning
|
A607143-10G
A607143-1G
A607143-250MG
A607143-5G
76992-098EA
482.71
USD
76992-098
76992-100
76992-102
76992-104
5-Chloro-1H-pyrazol-3-amine 98%
5-Chloro-1H-pyrazol-3-amine
Precaución: for Research Use Only
Formula:
C₃H₄ClN₃ MW: 117.537 g/mol |
MDL Number:
MFCD26518660 CAS Number: 916211-79-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...