tert-Butyl 2-(propan-2-ylidene)hydrazinecarboxylate 95%

Supplier: Ambeed
Warning

A607140-1G A607140-250MG
76992-094EA 40 USD
76992-094 76992-096
tert-Butyl 2-(propan-2-ylidene)hydrazinecarboxylate 95%
tert-Butyl 2-(propan-2-ylidene)hydrazinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₂
MDL Number: MFCD07636643
CAS Number: 16689-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR