2,4-Dichloro-5-fluoropyrimidine 98%

Supplier: Ambeed
Danger

Synonyms: 5-Fluoro-2,4-dichloropyrimidine

A606684-25G A606684-5G A606684-10G
77005-424EA 19.1 USD
77005-424 77005-426 77572-972
2,4-Dichloro-5-fluoropyrimidine 98%
2,4-Dichloro-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄HCl₂FN₂
MW: 166.97 g/mol
MDL Number: MFCD00233551
CAS Number: 2927-71-1
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR