2-[(4-Chlorophenyl)amino]acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: N-(4-Cl-Phenyl)-Gly-OH, 2-((4-Chlorophenyl)amino)acetic acid

A606483-100MG A606483-1G A606483-250MG
77018-010EA 44 USD
77018-010 77018-012 77018-014
2-[(4-Chlorophenyl)amino]acetic acid 95%
2-[(4-Chlorophenyl)amino]acetic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD00462901
CAS Number: 5465-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR