2,5-Difluoro-4-methoxybenzonitrile 97%

Supplier: Ambeed
Danger

A606479-10G A606479-1G A606479-25G A606479-5G
76992-080EA 93.36 USD
76992-080 76992-082 76992-084 76992-086
2,5-Difluoro-4-methoxybenzonitrile 97%
2,5-Difluoro-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂NO
MW: 169.13 g/mol
MDL Number: MFCD06246886
CAS Number: 1007605-44-8
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR