2,6-Difluoro-3-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A605334-1G A605334-5G
77005-422EA 28 USD
77005-422 77882-422
2,6-Difluoro-3-methylbenzaldehyde 98%
2,6-Difluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MDL Number: MFCD01631333
CAS Number: 261763-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR