N-Fmoc-N'-trityl-D-histidine 99% contains 3%CH2Cl2

Supplier: Ambeed
Warning

A605253-100G A605253-10G A605253-1G A605253-25G A605253-5G
77005-412EA 456 USD
77005-412 77005-414 77005-416 77005-418 77005-420
N-Fmoc-N'-trityl-D-histidine 99% contains 3%CH2Cl2
N-Fmoc-N'-trityl-D-histidine
Formula: C₄₀H₃₃N₃O₄
MDL Number: MFCD00077061
CAS Number: 135610-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR