Dipotassium hexahydroxoplatinate 98%

Supplier: Ambeed
Warning

A604954-100MG A604954-1G A604954-250MG A604954-5G
76992-068EA 44 USD
76992-068 76992-070 76992-072 76992-074
Dipotassium hexahydroxoplatinate 98%
Dipotassium hexahydroxoplatinate

Precaución: for Research Use Only

Formula: H₆K₂O₆Pt
MDL Number: MFCD00064671
CAS Number: 12285-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR