Tri-tert-butylmethanol 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 3-(Tert-butyl)-2,2,4,4-tetramethylpentan-3-ol

A604671-100MG A604671-250MG
77018-002EA 91.24 USD
77018-002 77684-476
Tri-tert-butylmethanol 98%, Ambeed.Inc
Tri-tert-butylmethanol

Precaución: For research use only.

Formula: C₁₃H₂₈O
MW: 200.361 g/mol
MDL Number: MFCD00204381
CAS Number: 41902-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR