6-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A603835-1G A603835-250MG
77018-000EA 102.91 USD
77018-000 77684-473
6-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
6-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: For research use only.

Formula: C₉H₁₀FN·ClH
MW: 187.642 g/mol
MDL Number: MFCD08437639
CAS Number: 799274-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR