6-Bromo-2-chloro-3-methylbenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-chloro-3-methylphenylboronic acid, (6-Bromo-2-chloro-3-methylphenyl)boronic acid

A602785-250MG A602785-5G
77005-402EA 5 USD
77005-402 77882-416
6-Bromo-2-chloro-3-methylbenzeneboronic acid 98%
6-Bromo-2-chloro-3-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrClO₂
MDL Number: MFCD09475910
CAS Number: 957120-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR