ML161 98%

Supplier: Ambeed
Warning

A601920-1MG A601920-250MG
77017-990EA 5 USD
77017-990 77017-992
ML161 98%
ML161

Precaución: for Research Use Only

Formula: C₁₇H₁₇BrN₂O₂
MDL Number: MFCD03063772
CAS Number: 423735-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR