5-Amino-2,4-di-tert-butylphenol 95%

Supplier: Ambeed
Warning

A601459-1G A601459-250MG A601459-5G
76992-054EA 55 USD
76992-054 76992-056 77891-189
5-Amino-2,4-di-tert-butylphenol 95%
5-Amino-2,4-di-tert-butylphenol

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO
MDL Number: MFCD11052607
CAS Number: 873055-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR