Pentane-1,5-diyl bis(3-(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)propanoate) dioxalate 97%

Supplier: Ambeed
Danger

A601200-5G A601200-1G
77035-116EA 80.63 USD
77035-116 77669-606
Pentane-1,5-diyl bis(3-(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)propanoate) dioxalate 97%
Pentane-1,5-diyl bis(3-(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)propanoate) dioxalate

Precaución: for Research Use Only

Formula: C₅₅H₇₀N₂O₂₀
MW: 1079.14 g/mol
MDL Number: MFCD11618009
CAS Number: 64228-78-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR