4-Bromonicotinonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-3-cyanopyridine

A600712-1G A600712-25G A600712-5G A600712-250MG
77005-388EA 47 USD
77005-388 77005-390 77005-392 77673-616
4-Bromonicotinonitrile 95%
4-Bromonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂
MW: 183.01
MDL Number: MFCD08686971
CAS Number: 154237-70-4
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR