1-Boc-3-(iodomethyl)azetidine 98%

Supplier: Ambeed
Warning

A600214-1G A600214-5G
77035-106EA 14 USD
77035-106 77035-108
1-Boc-3-(iodomethyl)azetidine 98%
1-Boc-3-(iodomethyl)azetidine

Precaución: for Research Use Only

Formula: C₉H₁₆INO₂
MDL Number: MFCD09951816
CAS Number: 253176-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR