(S)-tert-Butyl (6-oxopiperidin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A599732-100MG A599732-1G A599732-250MG A599732-5G
77035-098EA 20 USD
77035-098 77035-100 77035-102 77035-104
(S)-tert-Butyl (6-oxopiperidin-3-yl)carbamate 97%
(S)-tert-Butyl (6-oxopiperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MDL Number: MFCD13195282
CAS Number: 172913-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR