N-Boc-3-bromoaniline 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3-bromophenyl)carbamate

A598247-1G A598247-25G A598247-5G A598247-100G
77005-374EA 8 USD
77005-374 77005-376 77005-378 77882-409
N-Boc-3-bromoaniline 98%
N-Boc-3-bromoaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₂
MW: 272.1400 g/mol
MDL Number: MFCD01006618
CAS Number: 25216-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR