5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin 98%

Supplier: Ambeed
Danger

Synonyms: 4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl)tetrakis-Phenol, 5,10,15,20-Tetrakis(4-hydroxyphenyl)-21H,23H-porphine

A597516-100MG A597516-1G A597516-250MG A597516-5G
76992-038EA 14 USD
76992-038 76992-040 76992-042 76992-044
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin 98%
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin

Precaución: for Research Use Only

Formula: C₄₄H₃₀N₄O₄
MDL Number: MFCD00191501
CAS Number: 51094-17-8
UN: 3263
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR