4-(Trifluoromethoxy)benzenesulfonamide 97%

Supplier: Ambeed
Warning

A595194-100G A595194-10G A595194-25G A595194-5G
77005-344EA 109.28 USD
77005-344 77005-346 77005-348 77005-350
4-(Trifluoromethoxy)benzenesulfonamide 97%
4-(Trifluoromethoxy)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₆F₃NO₃S
MW: 241.19 g/mol
MDL Number: MFCD00798381
CAS Number: 1513-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR