1,3-Dimethylimidazolium methyl sulfate 98%

Supplier: Ambeed
Warning

A595145-1G A595145-25G A595145-5G
77017-968EA 7 USD
77017-968 77017-970 77017-972
1,3-Dimethylimidazolium methyl sulfate 98%
1,3-Dimethylimidazolium methyl sulphate

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O₄S
MDL Number: MFCD03453276
CAS Number: 97345-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR