5-(Methylsulfonyl)pyridine-3-boronic acid pinacol ester 97%

Supplier: Ambeed
Danger

A594948-1G A594948-250MG A594948-100MG A594948-5G
76992-028EA 195 USD
76992-028 77572-918 77594-812 77886-438
5-(Methylsulfonyl)pyridine-3-boronic acid pinacol ester 97%
5-(Methylsulfonyl)pyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₄S
MDL Number: MFCD12198145
CAS Number: 1206641-26-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR