3-Bromo-2-chloro-4-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-chloro-4-pyridinylboronic acid, (3-Bromo-2-chloropyridin-4-yl)boronic acid, 3-Bromo-2-chloropyridine-4-boronic acid

A593555-1G A593555-250MG A593555-5G
77035-078EA 57.29 USD
77035-078 77035-080 77035-082
3-Bromo-2-chloro-4-pyridineboronic acid 98%
3-Bromo-2-chloro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BBrClNO₂
MW: 236.26 g/mol
MDL Number: MFCD08458470
CAS Number: 1072944-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR